Order Entry
Denmark
ContactUsLinkComponent
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](4'-methoxyacetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](4'-methoxyacetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](4'-methoxyacetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together