Order Entry
Denmark
ContactUsLinkComponent
Protective hand creams, Handy Plus
Protective hand creams, Handy Plus
Catalog # 115-1377
Supplier:  Plum Safety
undefined
Protective hand creams, Handy Plus
Catalog # 115-1377
Supplier:  Plum Safety

Specifications

  • Description:
    Protective hand cream, Handy Plus, 700 ml bag-in-box
  • Pk:
    1

Specifications

About this item

Slightly perfumed skin care cream with regenerating effect for normal and dry skin.

  • Helps skin to restore natural barrier function
  • Quickly absorbs
  • For dispenser systems CombiPlum 0,5 L thread or plastic and MP2000
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904