Order Entry
Denmark
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ SPEC
Solid phase extraction cartridges, Bond Elut™ SPEC
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ SPEC
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ SPEC
Solid phase extraction cartridges, Bond Elut™ SPEC
25 Options Available Below
Product Details & Documents
SPEC SPE solid phase extraction cartridges available in standard straight barrel tube format, offering flexibility in sample size. Use on any standard vacuum manifold such as the Vac Elut 20 or SPS 24.
Product Family Options
SPEC SPE catridges
Product Information
  • Phase
    Pore size
    Capacity
    Bed weight
    Availability
    Price
  • VARIA5320320
    Straight barrel cartridge
    Straight barrel cartridge
    C18
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.570,00 kr
    3.570,00 kr
    1 * 100 Items
       3.570,00 kr
  • VARIA5320330
    Straight barrel cartridge
    Straight barrel cartridge
    C18
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.920,00 kr
    3.920,00 kr
    1 * 100 Items
       3.920,00 kr
  • VARIA5321920
    Straight barrel cartridge
    Straight barrel cartridge
    C18-AR
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.570,00 kr
    3.570,00 kr
    1 * 100 Items
       3.570,00 kr
  • VARIA5321930
    Straight barrel cartridge
    Straight barrel cartridge
    C18-AR
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.660,00 kr
    3.660,00 kr
    1 * 100 Items
       3.660,00 kr
  • VARIA5021935
    Straight barrel cartridge
    Straight barrel cartridge
    C18-AR
    70 Å
    10 ml
    35 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5022570
    Straight barrel cartridge
    Straight barrel cartridge
    C18-AR/MP3
    70 Å
    10 ml
    70 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 5.770,00 kr
    5.770,00 kr
    1 * 100 Items
       5.770,00 kr
  • VARIA5320130
    Straight barrel cartridge
    Straight barrel cartridge
    C2
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.920,00 kr
    3.920,00 kr
    1 * 100 Items
       3.920,00 kr
  • VARIA5320220
    Straight barrel cartridge
    Straight barrel cartridge
    C8
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.570,00 kr
    3.570,00 kr
    1 * 100 Items
       3.570,00 kr
  • VARIA532DAS
    Straight barrel cartridge
    Straight barrel cartridge
    DAS
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA532DAU
    Straight barrel cartridge
    Straight barrel cartridge
    DAU
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 2.780,00 kr
    2.780,00 kr
    1 * 100 Items
       2.780,00 kr
  • VARIA5321120
    Straight barrel cartridge
    Straight barrel cartridge
    MP1
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 4.740,00 kr
    4.740,00 kr
    1 * 100 Items
       4.740,00 kr
  • VARIA5321130
    Straight barrel cartridge
    Straight barrel cartridge
    MP1
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 5.070,00 kr
    5.070,00 kr
    1 * 100 Items
       5.070,00 kr
  • VARIA5021135
    Straight barrel cartridge
    Straight barrel cartridge
    MP1
    70 Å
    10 ml
    35 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5322020
    Straight barrel cartridge
    Straight barrel cartridge
    MP3
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 4.740,00 kr
    4.740,00 kr
    1 * 100 Items
       4.740,00 kr
  • VARIA5322030
    Straight barrel cartridge
    Straight barrel cartridge
    MP3
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 5.070,00 kr
    5.070,00 kr
    1 * 100 Items
       5.070,00 kr
  • VARIA5020735
    Straight barrel cartridge
    Straight barrel cartridge
    NH₂
    70 Å
    10 ml
    35 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5320720
    Straight barrel cartridge
    Straight barrel cartridge
    NH₂
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 3.890,00 kr
    3.890,00 kr
    1 * 100 Items
       3.890,00 kr
  • VARIA5020770
    Straight barrel cartridge
    Straight barrel cartridge
    NH₂
    70 Å
    10 ml
    70 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5321020
    Straight barrel cartridge
    Straight barrel cartridge
    Phenyl
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5321030
    Straight barrel cartridge
    Straight barrel cartridge
    Phenyl
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5320520
    Straight barrel cartridge
    Straight barrel cartridge
    SAX
    70 Å
    3 ml
    15 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
  • VARIA5320530
    Straight barrel cartridge
    Straight barrel cartridge
    SAX
    70 Å
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 4.330,00 kr
    4.330,00 kr
    1 * 100 Items
       4.330,00 kr
  • VARIA5020535
    Straight barrel cartridge
    Straight barrel cartridge
    SAX
    70 Å
    10 ml
    35 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items
SPEC disks, 47 mm
Product Information
  • Phase
    Pore size
    Capacity
    Bed weight
    Availability
    Price
  • Disk
    C8
    Log in for availability
    Log in for availability
    1 * 24 Items
    1 * 24 Items - 2.090,00 kr
    2.090,00 kr
    1 * 24 Items
       2.090,00 kr
  • Disk
    C18-AR
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 2.140,00 kr
    2.140,00 kr
    1 * 20 Items
       2.140,00 kr
96-well plates, SPEC
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, C2
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 0,00 kr
    0,00 kr
    1 * 1 Items
       0,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, C8
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.030,00 kr
    5.030,00 kr
    1 * 1 Items
       5.030,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, C18AR
    30 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.520,00 kr
    5.520,00 kr
    1 * 1 Items
       5.520,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, C18
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.030,00 kr
    5.030,00 kr
    1 * 1 Items
       5.030,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, NH2
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 0,00 kr
    0,00 kr
    1 * 1 Items
       0,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, phenyl
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 0,00 kr
    0,00 kr
    1 * 1 Items
       0,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, C18AR
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.030,00 kr
    5.030,00 kr
    1 * 1 Items
       5.030,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, DAU
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.360,00 kr
    5.360,00 kr
    1 * 1 Items
       5.360,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, SCX
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 0,00 kr
    0,00 kr
    1 * 1 Items
       0,00 kr
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • SPEC 96-well plates, SAX
    15 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 0,00 kr
    0,00 kr
    1 * 1 Items
       0,00 kr
Product Information
  • Catalog #
    Description
    Availability
    Price
  • SPEC 96 method development kit, with C2, C8, C18, C18AR, CN, MP1, MP3, and PH
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 5.980,00 kr
    5.980,00 kr
    1 * 1 Items
       5.980,00 kr
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Environmental disk manifold
    Environmental disk manifold
    VARIA712
    Log in for availability
    1 * 1 Items - 11.590,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Environmental disk holder, 47 mm
    Environmental disk holder, 47 mm
    VARIA713
    Log in for availability
    1 * 1 Items
Product Information
  • Catalog #
    Availability
    Price
  • Flask, 1 l, male 40/35 mm, ground glass fitting
    Flask, 1 l, male 40/35 mm, ground glass fitting
    VARIA714
    Log in for availability
    1 * 1 Items - 3.880,00 kr