Order Entry
Denmark
Orders LinkContactUsLinkComponent
SPE columns, CHROMABOND® C8
SPE columns, CHROMABOND® C8
Supplier:   MACHEREY-NAGEL
SPE columns, CHROMABOND® C8
Supplier:  MACHEREY-NAGEL
Certificates
SPE columns, CHROMABOND® C8
7 Options Available Below
Product Details & Documents
CHROMABOND® C8 is an octyl modified silica phase for SPE.
  • Slightly more polar than C18 phase
  • Hydrophobic and more pronounced secondary interactions with polar compounds
  • Suitable for slight nonpolar and polar substances
Recommended for polar pesticides and PCBs.
Specifications for Product Family
Specifications
  • Catalog No:
  • MANA730021
  • MANA730022
  • MANA730024
  • MANA730134
  • MANA730024G
  • MANA731808
  • MANA738021.100M
  • Carbon load:
  • 8%
  • 8%
  • 8%
  • 8%
  • 8%
  • 8%
  • Endcapped:
  • No
  • No
  • No
  • No
  • No
  • No
  • pH range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g
Specifications
Product Family Options
CHROMABOND® C8 polypropylene columns
Product Information
  • Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C8
    PP
    60 Å
    45 µm
    1 ml
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 Items
    1 * 100 Items - 1.960,00 kr
    1.960,00 kr
    1 * 100 Items
       1.960,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA730021
    Carbon load
    8%
    Endcapped
    No
    pH range
    2 - 8
    Surface area
    500 m²/g
  • C8
    PP
    60 Å
    45 µm
    3 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 1.520,00 kr
    1.520,00 kr
    1 * 50 Items
       1.520,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA730022
    Carbon load
    8%
    Endcapped
    No
    pH range
    2 - 8
    Surface area
    500 m²/g
  • C8
    PP
    60 Å
    45 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 Items
    1 * 30 Items - 1.150,00 kr
    1.150,00 kr
    1 * 30 Items
       1.150,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA730024
    Carbon load
    8%
    Endcapped
    No
    pH range
    2 - 8
    Surface area
    500 m²/g
  • C8
    PP
    60 Å
    45 µm
    6 ml
    1000 mg
    Log in for availability
    Log in for availability
    1 * 30 Items
    1 * 30 Items - 1.460,00 kr
    1.460,00 kr
    1 * 30 Items
       1.460,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA730134
    Carbon load
    8%
    Endcapped
    No
    pH range
    2 - 8
    Surface area
    500 m²/g
CHROMABOND® C8 glass columns
Product Information
  • Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C8
    Glass
    60 Å
    45 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 Items
    1 * 30 Items - 2.280,00 kr
    2.280,00 kr
    1 * 30 Items
       2.280,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA730024G
    Carbon load
    8%
    Endcapped
    No
    pH range
    2 - 8
    Surface area
    500 m²/g
CHROMAFIX® C8 cartridges
Product Information
  • Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C8
    Cartridge
    PP
    60 Å
    45 µm
    >210 mg
    Log in for availability
    Log in for availability
    1 * 50 Items
    1 * 50 Items - 1.950,00 kr
    1.950,00 kr
    1 * 50 Items
       1.950,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA731808
    Carbon load
    8%
    Endcapped
    No
    pH range
    2 - 8
    Surface area
    500 m²/g
CHROMABOND® MULTI 96 C8
Product Information
  • Phase
    Column Format
    Column hardware
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • C8
    60 Å
    45 µm
    96×100 mg
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 2.830,00 kr
    2.830,00 kr
    1 * 1 Items
       2.830,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    MANA738021.100M
    Carbon load
    Endcapped
    pH range
    Surface area