Order Entry
Denmark
ContactUsLinkComponent
Cryogenic dual storage vesssel, Arctic Express®
Cryogenic dual storage vesssel, Arctic Express®
Certificates
Cryogenic dual storage vesssel, Arctic Express®
Specifications
  • Catalog No:
  • BARNCK50922
  • BARNCK50920
  • BARNCK50921
  • Model:
  • Arctic Express Dual 28
  • Arctic Express Dual 10
  • Arctic Express Dual 19
  • Capacity:
  • 28 L
  • 10 L
  • 18,5 L
  • Daily evaporation:
  • 8 L
  • 3 L
  • 3 L
  • Int.Ø:
  • 97 mm
  • 50,8 mm
  • 56 mm
  • Ø×H:
  • 462×559 mm
  • 259×549 mm
  • 368×653 mm