Order Entry
Denmark
ContactUsLinkComponent
Wash bottles, narrow neck
Wash bottles, narrow neck
Catalog # KAUT76083
Leverandør:  Kautex
undefined
Wash bottles, narrow neck
Catalog # KAUT76083
Leverandør:  Kautex
Specifikationer
  • Pk:
    1
Specifikationer
Om denne vare
LDPE, translucent with LDPE spray cap.
  • Good chemical resistance
  • Heat resistant up to 80 °C
  • Leakproof
Economical, soft and very easy to squeeze. For use with general solutions such as water, soap, dilute acids and dilute alkalis. The tip can be cut off to increase the flow rate.