Order Entry
Denmark
Orders LinkContactUsLinkComponent
wells_lamont_sterile_gloves_955x590.jpg
Sterile Gloves
All Wells Lamont sterile glove liners are made and sterilized in the USA for high quality and quick shipping. These products have complete documentation and are trusted daily by end users in different applications.
Sterile Gloves | Avantor
sterile_gloves_low_400x495.jpg
Single use low level cut resistant Category III for particulates sterile glove liner.
sterile_gloves_mod_400x495.jpg
ANSI A2 cut resistant reusable Category II for particulates sterile glove liner.
SterilehighCut400x495.jpg
The M321 is a US made and sterilized glove liner that offers ANSI A4 cut resistance and tests Category III for particulates.