Order Entry
Denmark
Orders LinkContactUsLinkComponent
Glove-Liners-Hero-card955x590.jpg
Glove Liners
Wells Lamont Industrial has one of the most comprehensive selections of glove liners which can be worn underneath disposable gloves or by themselves. Options include low lint, touchscreen, antimicrobial, sterile, disposable, reusable, and cut resistant.
Wells Lamont Glove Liners | Avantor
Polyester-WireCut400x495.jpg
Cut resistant glove liner commonly used in cleanroom construction
The M281 glove liner offers ANSI 4 cut resistance while being rated at Category III for particulates. Highly reusable with great dexterity.
reusuable-nylon-liner400x495.jpg
Low lint highly reusuable nylon liner
The M555 liner is rated at Category II for particulates and is reusable up to 30 times.
Nylon-Glove-Liners400x495.jpg
Reusable glove liners for underneath disposable gloves
The M088 and M089 are great options when liners are needed underneath a disposable glove. Proudly made in the USA.
Cotton-Lisle-Inspection-Gloves400x495.jpg
Disposable glove liner for underneath disposable gloves
The Y6701 is a single use low cost disposable glove for use underneath disposable gloves.
Thin-glove-liner400x495.jpg
The M321 is a US Made and sterilized glove liner that offers ANSI A4 cut resistance and tests at Category III for particulates.
Thin-glove-liner400x495.jpg
The M121 glove liner offers ANSI 4 cut resistance and is highly reusable with great dexterity. Made with antimicrobial fibers.
Fiber-Sterile400x495.jpg
The M102 sterile liner offers ANSI Level 1 cut resistance and is a disposable product
Stretch-Glove-Liners400x495.jpg
The M104 sterile liner offers ANSI Level A2 cut resistance and can be reused up to 12 times.
Highly-reusableliners-400x495.jpg
Highly reusable nylon liners
Available in full finger (M005) and half finger (M006) these liners are reusable up to 30 times for a lower cost per use.
Spectra400x495.jpg
Cut Resistant liner
The M214 offers ANSI Level 2 cut resistance, is light weight, and highly reusable.
Kevlar-cut-resistant-liner-400x495.jpg
Light weight Kevlar liner offers ANSI Level 2 cut resistance.
Nylon-GloveAutofit400x495.jpg
Medical nylon liners
Full finger and half finger medical nylon liners packaged in boxes of 25 gloves. Reusuable up to 20 times.
Stretch-Glove-Liners400x495.jpg
The M114 liner offers ANSI A2 cut resistance and is reusable up to 12 times