Order Entry
Denmark
Orders LinkContactUsLinkComponent
Wells Lamont Cleanroom Gloves | Avantor
Hero_sf_808.jpg
Cleanroom Gloves
Cleanroom glove liners from Wells Lamont are used everyday to protect people and their environment. These products are made in the USA for high quality and reliable availability.
Block1_sf_808.jpg
The M281 glove liner offers ANSI 4 cut resistance while being rated at Category III for particulates. Highly reusable with great dexterity.
Block2_sf_808.jpg
The M555 liner is rated at Category II for particulates and is reusable up to 30 times.
Block3_sf_808.jpg
The M321 is a US Made and sterilized glove liner that offers ANSI A4 cut resistance and tests at Category III for particulates.
M102Spectecspectra_400x495.jpg
Single use low level cut resistant Category III for particulates sterile glove liner.
M104Spectecspectra_400x495.jpg
ANSI A2 cut resistant reusable Category II for particulates sterile glove liner.
M214Spectecspectra_400x495.jpg
Category II cleanroom A2 cut resistant liner.
Silver_Talon_400x495.jpg
Category III cleanroom ANSI A7 cut resistant glove.