Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hero_sf_794.jpg
PIG® HazMat Spill Kits
The only spill kits packed full of PIG HazMat Absorbents!
PIG HazMat Spill Kits | Avantor
WORLD-CLASS ABSORBENTS
Absorb the widest range of acids, bases and unknown liquids, even those with high concentrations like 98% sulfuric acid and 30% sodium hydroxide.
VARIETY OF CONTAINERS
Choices range from portable grab-and-go to mobile carts to overpack salvage drums.
PACKED FOR FAST RESPONSE
Kits are packed in the order contents are needed for fast response and access.
High_visibility_sf_794.jpg
PIG® HazMat Spill Kit in High-Visibility Bag
Absorbs up to 9 gal., Absorbs Acids; Bases; Unknowns PIG Chemical Absorbents in an easy to see, fast to find, hi-viz bag.
High-Visibility_Caddy_sf_794.jpg
Absorbs up to 14 gal., Absorbs Acids, Bases, Unknowns .Fully stocked, wheeled mobile cart features swing out front doors for easy access.
Block3_sf_794.jpg
Absorbs up to 63 gal., Absorbs Acids; Bases; Unknowns, UN1H2/X113/S Strong enough to earn a UN rating for shipping chemical spill cleanup waste. 
Block4_sf_794.jpg
Absorbs up to 11 gal., Absorbs Acids; Bases; Unknowns, UN1H2/X113/S Strong enough to earn a UN rating for shipping chemical spill cleanup waste.
Block6_sf_794.jpg
VWR Lab Spill Kit
Have your lab ready with this compact Kit. It’s packed with HAZ-MAT absorbents and PPE to safely contain and soak up small spills of chemicals.