Order Entry
Denmark
Orders LinkContactUsLinkComponent
Chromasolv540-390.jpg
Chromasolv™ and Burdick & Jackson™ High Purity Solvents
Choose from more than 100 high purity solvents, including Honeywell Chromasolv™ and Burdick & Jackson™ solvents, and Honeywell LabReady™ blends.  All solvents have low impurity levels and high lot-to-lot consistency which are tailored to your specific application needs.
The Honeywell high purity solvents are suitable for a variety of applications including liquid and gas chromatography (LC and GC), spectrophotometry, pesticide residue analysis, organic synthesis and combinatorial chemistry.
Honeywell Research Chemicals has over 200 years of expertise in high purity solvents, produced in two ISO 9001 certified production plants in the USA and Germany.
Product selection
Chromasolv100.jpg
Chromasolv™ solvents are thoroughly tested to ensure suitability for general High-Performance Liquid Chromatography (HPLC) or HPLC gradient analysis.
Brand-Solvents100.jpg
Solvents for a variety of applications including LC, GC, spectrophotometry, pesticide residue analysis, organic synthesis and combinatorial chemistry
ChromasolvGC100.jpg
Solvents specifically developed for GC and GC-headspace analysis including analysis of volatile organics, pesticide residues, dioxins, furans and PCB.
GC2-Solvents100.jpg
Selection of solvents for environmental analysis such as capillary GC applications requiring trace analysis at or below the part-per-billion level.
Chromasolv-LC-MS100.jpg
Solvents for the most demanding applications undergo several tests to ensure highest quality for reliable LC-MS or UHPLC-MS analysis.