Order Entry
Denmark
Orders LinkContactUsLinkComponent
DuPont_Tychem_-products_hero_955x590.jpg
DuPont™ Tychem® garments
DuPont™ Tychem® garments | Avantor
DuPont™ Tychem® garments for lightweight protection from chemical and biological hazards. Not all deadly threats come in the form of fires or weapons. For comprehensive protection against low levels of dangerous chemicals, to toxic gases and biological hazards, industrial workers rely on garments made of DuPont Tychem®. Tychem® is an innovative fabric range comprising multiple barrier layers that help provide protection from chemical and biological hazards. Laminated to Tyvek® for strength, it offers a robust solution with a high level of chemical barrier at uncharacteristically low weight.
Learn more
Tychem_6000_F_FaceSeal_400x495.jpg
Tight design combined with trusted Tychem® protection. DuPont Tychem® 6000 F FaceSeal. Hooded coverall with rubber seal on hood for tight fit around respirator.
TYC_C_SOCKS_HOL_400x495.jpg
Tychem® 2000 C
Tychem® 2000 C garments utilise the strength of Tyvek®, and a polymeric barrier coating, to offer good permeation barrier protection against a wide range of inorganic chemicals and biological hazards (even under pressure).
Video
Literature