Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cardinal Health Sterile Gloves | Avantor
Cardinal Health Sterile Gloves
Product selection
Sterile_latex.jpg
CR100® Sterile Latex Gloves
Versatile, high-quality latex gloves can be used in critical environments that demand stringent aseptic procedures. ISO 4
DurapreneCPSyntheticGloves.png
Duraprene™CP Synthetic Gloves
Nitrile and neoprene blend that is latex-free. Hand specific with beaded cuffs as well as powder-free. ISO 5
C100BTLatexLaboratoryGloves.png
CP100BT Latex Laboratory Gloves
Sterile, clean-processed gloves feature a comfortable flex-and-stretch design and low particulate levels for minimized contamination of controlled environments. ISO 5
EsteemCPSterileSyntheticGloves.png
Esteem® CP Sterile Synthetic Gloves, Cardinal Health®
Hand-specific, powder-free clean process gloves are made of polyisoprene synthetic polymer for a fit, feel, and performance that is virtually indistinguishable from latex. ISO 6
ProtegrityCPLatexGloves.png
Protegrity® CP Latex Gloves
Triple-layer, powder-free, clean process gloves. Has exceptional grip, outstanding tactile properties, and enhanced comfort. ISO 6