Order Entry
Denmark
Orders LinkContactUsLinkComponent
CardinalHealthNonSterileGloves-955x690.jpg
Cardinal Health Non-Sterile Gloves
Cardinal Health Non-Sterile Gloves | Avantor
Product Selection
CP100NitrileGloves-400x495.jpg
Clean-processed nitrile gloves offer excellent strength and a comfortable fit. ESD properties and 29.2cm (111/2") length make gloves ideal for many cleanroom applications. ISO 6
CP100BTAmbiLatex-400x495.jpg
CP 100BT Ambi Latex
Ambidextrous gloves feature a beaded cuff to minimize rolldown and make donning easy. Fully textured palm and fingers provide a secure grip. Clean-processed, powder-free. ISO 6