Orders LinkContactUsLinkComponent
[EN]BUFFER SOLUTION PH 4.00 (25°C) 1 * 1 L
[EN]BUFFER SOLUTION PH 4.00 (25°C) 1 * 1 L
Katalog # LOVI721252
New Product
Leverandør:   Lovibond
[EN]BUFFER SOLUTION PH 4.00 (25°C) 1 * 1 L
Katalog # LOVI721252
Leverandør:  Lovibond
New Product
[EN]BUFFER SOLUTION PH 4.00 (25°C) 1 * 1 L
Price
418,00 kr
Log ind for tilgængelighed
1 * 1 L