[EN]100F-CTC-GT-5/0.47C SYRINGE CALIBRAT 1 * 1 Items
Katalog # SGEA005700CAL
Leverandør: Trajan Scientific and Medical
New Product
[EN]100F-CTC-GT-5/0.47C SYRINGE CALIBRAT 1 * 1 Items
Katalog # SGEA005700CAL
Leverandør: Trajan Scientific and Medical
New Product
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 5 µm
Leverandør: Avantor
Tilbehør til SCAT SafetyCaps
Leverandør: S.C.A.T.
Stopcocks, miniature
Leverandør: Bohlender
N NEEDLE (16/51/3) 1 * 6 Items
Leverandør: HAMILTON BONADUZ
SKRUELÅG U/HUL -butyl/PFTE liner- St. 24-400 - Til BROW150907-30 ml, Brow150908-40ml og Brow150909-60 ml 1 * 100 Items
Leverandør: BROWN CHROMATOGRAPHY SUPPLIES
HPLC columns, Hypersil™ ODS-2 C18
Leverandør: Thermo Fisher Scientific
[EN]DISCOVERY DSI-70 PK/54 3ML TUBE 500M 1 * 54 Items
Leverandør: Merck
Safety waste caps and ventilation kit, SymLine®
Leverandør: S.C.A.T.
Screw caps for screw neck vials, N 9
Leverandør: MACHEREY-NAGEL
Syringes for gas chromatography autosamplers, CTC PAL® GC-xt and Combi PAL-xt liquid mode
Leverandør: HAMILTON BONADUZ
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Leverandør: PerkinElmer
Tubing connectors Y, b.safe
Leverandør: Bohlender