[EN]PUL NOMINAL MW 342 DA 0.1 G 1 * 1 Items
Katalog # AGLSPSS-PUL342
Leverandør: AGILENT
New Product
Frequently Bought Together
VWR® AVS TITRINORM, bufferopløsninger, 20 °C
Leverandør: VWR Chemicals
VWR®, Conductivity standard solutions
Leverandør: VWR Chemicals
Nitrogen (total) CRM, Standard Solutions, Supelco®
Leverandør: Merck
Kaliumchlorid 3 mol/l (3 N) in aqueous solution, Supelco®
Leverandør: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Leverandør: Merck
Nitrat 1.000 mg/l i vand (af NaNO<sub>3</sub>) Certipur®, Supelco®
Leverandør: Merck
pH buffer solutions
Leverandør: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Leverandør: Merck
Saltsyre 0.01 mol/l (0.01 N) Reag. Ph. Eur. 1043504
Leverandør: VWR Chemicals
di-Natrium oxalat, Certipur®, Supelco®
Leverandør: Merck
VWR®, Primary colour solutions, Reagent Ph. Eur.
Leverandør: VWR Chemicals
Ledningsevnestandarder
Leverandør: VWR Chemicals
pH buffer solutions
Leverandør: Hach