Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant MCP2 (from E. coli)

Human Recombinant MCP2 (from E. coli)

Katalog # ABCAAB316061-500UG
New Product
Leverandør:   Abcam
Human Recombinant MCP2 (from E. coli)
Katalog # ABCAAB316061-500UG
Leverandør:  Abcam
New Product
CAS nummer:  
Human Recombinant MCP2 (from E. coli)
Human Recombinant MCP2 (from E. coli)
CAS nummer:  
Price
14.650,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human CCL8 protein is a Human Fragment protein, in the 24 to 99 aa range, expressed in Escherichia coli, with >95% purity and <1 EU/µg endotoxin level.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    MCP2
  • Protein synonyms:
    SCYA10|SCYA8|CCL8|C-C motif chemokine 8|HC14|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|Small-inducible cytokine A8|MCP-2
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    Ambient - Can ship with ice
  • Cat. No.:
    ABCAAB316061-500UG
  • Protein/peptide type:
    Recombinant
  • Formulering:
    Constituents: 91,99% PBS, 8% Trehalose
  • Endotoksinindhold:
    <1 EU/µg
  • Renhed:
    >95%
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.