Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Human Recombinant MCP2 (from E. coli)

Catalog # ABCAAB316061-100UG
New Product
Supplier:   Abcam
Human Recombinant MCP2 (from E. coli)
Catalog # ABCAAB316061-100UG
Supplier:  Abcam
New Product
CAS Number:  
Human Recombinant MCP2 (from E. coli)
Human Recombinant MCP2 (from E. coli)
CAS Number:  
Price
3.690,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Recombinant Human CCL8 protein is a Human Fragment protein, in the 24 to 99 aa range, expressed in Escherichia coli, with >95% purity and <1 EU/µg endotoxin level.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    MCP2
  • Protein synonyms:
    SCYA10|SCYA8|CCL8|C-C motif chemokine 8|HC14|Monocyte chemoattractant protein 2|Monocyte chemotactic protein 2|Small-inducible cytokine A8|MCP-2
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    N
  • Shipping temperature:
    Ambient - Can ship with ice
  • Cat. No.:
    ABCAAB316061-100UG
  • Protein/peptide type:
    Recombinant
  • Formulering:
    Constituents: 91,99% PBS, 8% Trehalose
  • Endotoksinindhold:
    <1 EU/µg
  • Renhed:
    >95%
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –20 °C

Specifications