Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant SCF (from E. coli)

Human Recombinant SCF (from E. coli)

Katalog # ABCAAB316045-500UG
New Product
Leverandør:   Abcam
Human Recombinant SCF (from E. coli)
Katalog # ABCAAB316045-500UG
Leverandør:  Abcam
New Product
CAS nummer:  
Human Recombinant SCF (from E. coli)
Human Recombinant SCF (from E. coli)
CAS nummer:  
Price
9.680,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant Human SCF (Active) protein is a Human Fragment protein, in the 26 to 189 aa range, expressed in Escherichia coli, with >95% purity and <1 EU/µg endotoxin level.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    SCF
  • Protein synonyms:
    MGF|KITLG|Kit ligand|Mast cell growth factor|Stem cell factor|c-Kit ligand
  • Biologisk aktivitet:
    Bioactive
  • Dyrefri:
    N
  • Shipping temperature:
    Ambient - can ship with ice
  • Cat. No.:
    ABCAAB316045-500UG
  • Protein/peptide type:
    Recombinant
  • Formulering:
    Constituents: 91,99% PBS, 8% Trehalose
  • Endotoksinindhold:
    <1 EU/µg
  • Renhed:
    >95%
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at −20 °C

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.