Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant KCNE2 (from E. coli)

Human Recombinant KCNE2 (from E. coli)

Katalog # ABCAAB314528-20UG
New Product
Leverandør:   Abcam
Human Recombinant KCNE2 (from E. coli)
Katalog # ABCAAB314528-20UG
Leverandør:  Abcam
New Product
CAS nummer:  
Human Recombinant KCNE2 (from E. coli)
Human Recombinant KCNE2 (from E. coli)
CAS nummer:  
Price
15.980,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Recombinant human KCNE2 protein (His tag) is a Human Full Length protein, in the 1 to 123 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE.

Advarsel: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifikationer

Specifikationer

  • Konjugation:
    Unconjugated
  • Form:
    Lyophilised
  • Protein/peptide name:
    KCNE2
  • Protein synonyms:
    Potassium voltage-gated channel subfamily E member 2|MinK-related peptide 1|Minimum potassium ion channel-related peptide 1|Potassium channel subunit beta MiRP1|MiRP1|KCNE2
  • Biologisk aktivitet:
    Standard
  • Dyrefri:
    Y
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB314528-20UG
  • Protein/peptide type:
    Recombinant
  • Formulering:
    pH: 7,4 - 8 Constituents: 6% Trehalose, 0,87% Sodium chloride, 0,24% Tris, 0,05% dodecyl 2-(trimethylazaniumyl)ethyl phosphate
  • Renhed:
    >90%
  • Tested applications:
    SDS-PAGE
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Store at –20 °C

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.