Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human IgM ELISA Kit

Human IgM ELISA Kit

Human IgM ELISA kit, 384 tests
Katalog # ABCAAB214568-1X384
New Product
Leverandør:   Abcam
Human IgM ELISA Kit
Katalog # ABCAAB214568-1X384
Leverandør:  Abcam
New Product
CAS nummer:  
Human IgM ELISA Kit
Human IgM ELISA Kit

Human IgM ELISA kit, 384 tests

CAS nummer:  
Price
15.010,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Human IgM ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IgM with a sensitivity of 50 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput. - Cited in over 5 citations.

Specifikationer

Specifikationer

  • Beskrivelse:
    Human IgM ELISA kit
  • Test pr. sæt:
    384 tests
  • Sensitivitet:
    50 pg/ml
  • Assay varighed:
    One step
  • Primary antibody reactivity:
    Human
  • Detektionsområde:
    390,63 - 25000 pg/ml
  • Prøvetype:
    Saliva, Heparin Plasma, Citrate plasma, Cell culture supernatant, Milk, Serum, EDTA Plasma
  • Time to Results:
    1 h 30 min
  • Analysetype:
    Sandwich
  • Cat. No.:
    ABCAAB214568-1X384
  • Format:
    Pre-coated
  • Target protein:
    IgM
  • Detektionsmetode:
    Colorimetric
  • Opbevaringstemperatur:
    +4 °C
  • Regulatory status:
    RUO

Specifikationer