Temperature Probes for Heating Mantle Controller
Temperature probes SS400, standard version, 4,8×380 mm
Katalog # WITGDH.WHP503050
New Product
Leverandør: witeg Labortechnik
Temperature Probes for Heating Mantle Controller
Katalog # WITGDH.WHP503050
Leverandør: witeg Labortechnik
New Product
Gå til:
Product Details & Documents
Temperature probe for use with heating mantle controller WHM-C10D.
Specifikationer
- Beskrivelse:Temperature probes SS400, standard version
- Seq:2
- Ø×L:4,8×380 mm