Sodium Decanoate 98%
Bottle
Catalog # APOSOR1015017-500G
Supplier: Apollo Scientific
Gå til:
Product Details & Documents
Usage: Organic building block/reagent.
- CAS nummer:1002-62-6
- Formel:C₁₀H₁₉NaO₂
- MDL nummer:MFCD00066453
- Molecular Weight:194,25 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle



