Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sodium Decanoate 98%

Sodium Decanoate 98%

Bottle
Katalog # APOSOR1015017-100G
Leverandør:   Apollo Scientific
Sodium Decanoate 98%
Katalog # APOSOR1015017-100G
Leverandør:  Apollo Scientific
CAS nummer:  1002-62-6
Sodium Decanoate 98%

Bottle

CAS nummer:  1002-62-6
Price
615,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

  • CAS nummer:1002-62-6
  • Formel:C₁₀H₁₉NaO₂
  • MDL nummer:MFCD00066453
  • Molecular Weight:194,25 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.