Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Nonadecane 99%

Bottle
Catalog # APOSOR1014448-500G
Supplier:   Apollo Scientific
Nonadecane 99%
Catalog # APOSOR1014448-500G
Supplier:  Apollo Scientific
CAS Number:  629-92-5
Nonadecane 99%

Bottle

CAS Number:  629-92-5
Price
5.460,00 kr
Log in for availability
1 * 500 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonyms:  N-nonadecane
  • CAS Number:629-92-5
  • Formula:C₁₉H₄₀
  • MDL Number:MFCD00009012
  • Molecular Weight:268,52 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications