Order Entry
Denmark
Orders LinkContactUsLinkComponent
Nonadecane 99%

Nonadecane 99%

Bottle
Katalog # APOSOR1014448-100G
Leverandør:   Apollo Scientific
Nonadecane 99%
Katalog # APOSOR1014448-100G
Leverandør:  Apollo Scientific
CAS nummer:  629-92-5
Nonadecane 99%

Bottle

CAS nummer:  629-92-5
Price
1.220,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

Synonymer:  N-nonadecane
  • CAS nummer:629-92-5
  • Formel:C₁₉H₄₀
  • MDL nummer:MFCD00009012
  • Molecular Weight:268,52 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.