Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3-Hydroxyazetidine 98%

3-Hydroxyazetidine 98%

Bottle
Katalog # APOSOR301160-10G
Irriterende
Leverandør:   Apollo Scientific
3-Hydroxyazetidine 98%
Katalog # APOSOR301160-10G
Leverandør:  Apollo Scientific
CAS nummer:  45347-82-8
3-Hydroxyazetidine 98%

Bottle

CAS nummer:  45347-82-8
Price
5.940,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonymer:  3-Azetidinol, Azetidin-3-ol
  • CAS nummer:45347-82-8
  • Formel:C₃H₇NO
  • MDL nummer:MFCD03695446
  • Molecular Weight:73,09377 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.