ContactUsLinkComponent
~OBJEKTGLAS 103x76,3±0,3 mm, thickness 1 mm, white glass, cut edges, without frosted end, paper-interleaved, in boxes of 50 pcs. MARIENFELD SUPERIOR 1 * 50 Items
Katalog # MARI152310309
Leverandør:  MARIENFELD
New Product
~OBJEKTGLAS 103x76,3±0,3 mm, thickness 1 mm, white glass, cut edges, without frosted end, paper-interleaved, in boxes of 50 pcs. MARIENFELD SUPERIOR 1 * 50 Items
Katalog # MARI152310309
Leverandør:  MARIENFELD