Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
3-Hydroxy-5-methylbenzonitrile 98%

3-Hydroxy-5-methylbenzonitrile 98%

Bottle
Katalog # APOSOR928299-5G
New Product
Leverandør:   Apollo Scientific
3-Hydroxy-5-methylbenzonitrile 98%
Katalog # APOSOR928299-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  95658-81-4
3-Hydroxy-5-methylbenzonitrile 98%

Bottle

CAS nummer:  95658-81-4
Price
3.990,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
  • CAS nummer:95658-81-4
  • Formel:C8H7NO
  • MDL nummer:MFCD16999137
  • Molecular Weight:133.1500 g/mol
  • Opbevaringstemperatur:Stue temperatur
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.