4-Ethylpyrocatechol 97%
Bottle
Catalog # APOSOR925237-250MG
New Product
Supplier: Apollo Scientific
4-Ethylpyrocatechol 97%
Catalog # APOSOR925237-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 1124-39-6
Jump to:
Product Details & Documents
Synonyms: 4-Ethylbenzene-1,2-diol, 4-Ethylcatechol
- CAS Number:1124-39-6
- Formula:C₈H₁₀O₂
- MDL Number:MFCD00015847
- Molecular Weight:138.17 g/mol
- Storage Temperature:Stue temperatur
Specifications
Specifications
- Pakning:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



