Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Methyl Oleate mixture of isomers 99%

Bottle
Katalog # APOSOR1013872-100G
Irriterende
Leverandør:   Apollo Scientific
Methyl Oleate mixture of isomers 99%
Katalog # APOSOR1013872-100G
Leverandør:  Apollo Scientific
CAS nummer:  112-62-9
Methyl Oleate mixture of isomers 99%

Bottle

CAS nummer:  112-62-9
Price
187,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent.

  • Store at −20 °C
Synonymer:  cis-Oleic Acid Methyl Ester, Oleic acid methyl ester
  • CAS nummer:112-62-9
  • Formel:C₁₉H₃₆O₂
  • MDL nummer:MFCD00009578
  • Molecular Weight:296,49 g/mol
  • Opbevaringstemperatur:Fryser
  • EINECS:203-992-5
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer