Orders LinkContactUsLinkComponent
Creatinin 95+%
Creatinin 95+%
Bottle
  APOSOR905165-25G
New Product
Creatinin 95+%
  APOSOR905165-25G
New Product
 :  60-27-5
Creatinin 95+%
Bottle
 :  60-27-5
165,00 kr
1 * 25 g
 :  2-Amino-1-methyl-1,5-dihydro-4H-imidazol-4-one
  •  :60-27-5
  •  :C₄H₇N₃O
  •  :MFCD00059730
  •  :113.12 g/mol
  •  :Stue temperatur
  • Pakning:
    Bottle