Orders LinkContactUsLinkComponent
Accessories for Corning CellSTACK culture chambers
CellSTACK 10 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
Katalog # CORN11762
Leverandør:   Corning
Accessories for Corning CellSTACK culture chambers
Katalog # CORN11762
Leverandør:  Corning
Accessories for Corning CellSTACK culture chambers
Accessories for Corning CellSTACK culture chambers
CellSTACK 10 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
Price
14.490,00 kr
Log ind for tilgængelighed
1 * 4 Items
Specifikationer
  • Beskrivelse:
    CellSTACK 10 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
  • Seq:
    4