Accessories for Corning CellSTACK culture chambers
CellSTACK 5 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
Katalog # CORN11761
Leverandør: Corning
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:CellSTACK 5 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mm
- Seq:6