Cryo-Grip® Waterproof Gloves
Product Details & Documents
Waterproof Cryo-Grip® gloves offer improved grip on small objects while protecting hands and arms when working in hasardous, ultra-cold environments.
- Maximum thermal protection
- Lightweight and comfortable
- Excellent dexterity
- Breathable and stay-dry lining
Gloves are designed to provide a high level of thermal protection where the exposure to cryogenic fluids might exist. Made from state-of-the-art materials, these waterproof cryogenic gloves are made to wick away moisture and keep wearer hands dry, to work in comfort for extended periods of time.
Where other gripping materials crack and break in ultra-low environments, the innovative Cryo-Grip® fabric will stay flexible, even in extreme conditions. The multi-layered construction allows for a maximum level of grip, dexterity, flexibility, and thermal protection - essential features when function is important and safety is critical.
Certifications: Cat III. EN 420. EN 511. EN 388.
Specifications
- Handske størrelse:M
- Farve:blå
- Længde:435 - 495 mm
- Seq:11
- Cat. No.:TEMPCGEBMWP
- Ambidextrous/Hånd-specifik:Hånd-specifik
- Engangs/Genbrugelig:Genanvendelig