Orders LinkContactUsLinkComponent
NEXTFLEX™ Poly(A) Beads 2.0
NEXTFLEX™ Poly(A) Beads 2.0
NEXTFLEX™ Poly(A) beads 2.0, 48
Catalog # BIOONOVA-512992
Marketsource
Supplier:   Revvity
NEXTFLEX™ Poly(A) Beads 2.0
Catalog # BIOONOVA-512992
Supplier:  Revvity
Marketsource
CAS Number:  
NEXTFLEX™ Poly(A) Beads 2.0
NEXTFLEX™ Poly(A) Beads 2.0
NEXTFLEX™ Poly(A) beads 2.0, 48
CAS Number:  
Price
1.371,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
NEXTFLEX™ poly(A) beads 2.0 kits offer a convenient method for obtaining pure, intact mRNA upstream of RNA-sequence library prep with improved mRNA yields, low rRNA contamination and a simple protocol.
  • 10 ng to 5 µg total starting RNA Magnetic bead-based protocol No organic solvents or precipitation step required Optimized for use with 10 ng to 5 µg of total RNA as starting material Versions are available for automated and manual isolation of the mRNA transcriptome Automated on the Sciclone® G3 NGSx and Zephyr® G3 NGS workstations
Several approaches have been used to remove ribosomal RNA (rRNA) from total RNA samples for RNA-sequence library preparation. Removal of rRNA avoids the waste of reagents and bioinformatics resources to sequence and align ~85% of total RNA comprising rRNA (which is generally not a target of interest in NGS sequencing experiments). Revvity’s NEXTFLEX™ poly(A) beads 2.0 kit offers an easy and cost-effective method for removing rRNA in RNA-sequence libraries. This product takes advantage of the tract of adenosine residues present at the 3’ ends of a vast majority of protein-coding mRNAs. Hybridization of these poly(A) tails to tracts of complementary thymidines ('oligo dT') immobilised on solid supports allows the retrieval of poly(A)+ RNAs by recovering the solid support along with the hybridized RNA complexes.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
Specifications
  • Beskrivelse:
    NEXTFLEX™ Poly(A) beads 2.0
  • No. of reactions:
    48
Specifications