Skalpelblade
No. 24
Supplier: SWANN MORTON
Jump to:
Product Details & Documents
Kulstofstål.
- Available in sterile individual foil packs or non sterile foil packs of five
- Quick and easy to assemble
Specifications
- Model:No. 24
- Seq:35