Skalpelblade
No. 22
Leverandør: SWANN MORTON
Gå til:
Product Details & Documents
Kulstofstål.
- Available in sterile individual foil packs or non sterile foil packs of five
- Quick and easy to assemble
Specifikationer
- Model:No. 22
- Seq:31
Kulstofstål.