Orders LinkContactUsLinkComponent
Skalpelblade
No. 21
Katalog # SWAN1107
Leverandør:   SWANN MORTON
Skalpelblade
Katalog # SWAN1107
Leverandør:  SWANN MORTON
Skalpelblade
Skalpelblade
No. 21
Price
389,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Kulstofstål.
  • Available in sterile individual foil packs or non sterile foil packs of five
  • Quick and easy to assemble
Specifikationer
  • Model:
    No. 21
  • Seq:
    29