Orders LinkContactUsLinkComponent
Skalpelblade
Skalpelblade
No. 11P
Catalog # SWAN1191
Supplier:   SWANN MORTON
Skalpelblade
Catalog # SWAN1191
Supplier:  SWANN MORTON
Skalpelblade
Skalpelblade
No. 11P
Price
349,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
Kulstofstål.
  • Available in sterile individual foil packs or non sterile foil packs of five
  • Quick and easy to assemble
Specifications
  • Model:
    No. 11P
  • Seq:
    21