Orders LinkContactUsLinkComponent
Antibiotic Sensitivity Discs
Antibiotic Sensitivity Discs
Clarithromycin, 15 mcg, 50
Katalog # HIMESD192-5X50DS
Leverandør:   HIMEDIA
Antibiotic Sensitivity Discs
Katalog # HIMESD192-5X50DS
Leverandør:  HIMEDIA
CAS nummer:  
Antibiotic Sensitivity Discs
Clarithromycin, 15 mcg, 50
CAS nummer:  
Price
145,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Specifikationer
Specifikationer
  • Pakn:
    50
  • Beskrivelse:
    Clarithromycin, 15 mcg