Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Waste Containers, PE

Waste Containers, PE

Container, 265W×400H mm, 10000 ml
Katalog # METR6.1621.000
Leverandør:   METROHM
Waste Containers, PE
Katalog # METR6.1621.000
Leverandør:  METROHM
Waste Containers, PE

Container, 265W×400H mm, 10000 ml

Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

This is a rinsing or waste container in automated systems.

  • Made of PE
Specifikationer
  • Kapacitet:
    10000 ml
  • Beskrivelse:
    Container, 265W×400H mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.