Order Entry
Denmark
Orders LinkContactUsLinkComponent
Multi-Cassette (Urine)
Multi-Cassette (Urine)
10 parameter urine multi-cassette, AMP/BAR/BZO/COC/MET/MDMA/MOP/MTD/TCA/THC
Katalog # SUREG10MC2300
Leverandør:   SURESCREEN
Multi-Cassette (Urine)
Katalog # SUREG10MC2300
Leverandør:  SURESCREEN
Multi-Cassette (Urine)
10 parameter urine multi-cassette, AMP/BAR/BZO/COC/MET/MDMA/MOP/MTD/TCA/THC
Price
95,60 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
The Multi-Cassette only requires a small amount of urine to perform the test, with test results available at 5 minutes. This multiline format test comes with a pipette for an easy-to-use process.
  • Multi-drug test incorporated
  • Suitable for low-volume sample
  • Test up to 12 parameters simultaneously
Specifikationer
  • Beskrivelse:
    10 parameter urine multi-cassette
  • Analyser:
    AMP/BAR/BZO/COC/MET/MDMA/MOP/MTD/TCA/THC