Multi-Cassette (Urine)
10 parameter urine multi-cassette, AMP/BAR/BZO/COC/MET/MDMA/MOP/MTD/TCA/THC
Katalog # SUREG10MC2300
Leverandør: SURESCREEN
Gå til:
Product Details & Documents
The Multi-Cassette only requires a small amount of urine to perform the test, with test results available at 5 minutes. This multiline format test comes with a pipette for an easy-to-use process.
- Multi-drug test incorporated
- Suitable for low-volume sample
- Test up to 12 parameters simultaneously
Specifikationer
- Beskrivelse:10 parameter urine multi-cassette
- Analyser:AMP/BAR/BZO/COC/MET/MDMA/MOP/MTD/TCA/THC