Orders LinkContactUsLinkComponent
Axygen® LTS Compatible Pipette Tips
LTS compatible, extended length, 1000 µl, -, -, 98,58 mm, Clear, 8 racks, 96 each
  613-7409
 :   Corning
Axygen® LTS Compatible Pipette Tips
  613-7409
 :  Corning
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
LTS compatible, extended length, 1000 µl, -, -, 98,58 mm, Clear, 8 racks, 96 each
473,00 kr
1 * 768 Items
The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.
 : ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
  • Pakket:
    8 racks, 96 each
  • Steril:
    -
  • Volumen:
    1000 µl
  • Længde:
    98,58 mm
  • Seq:
    09
  • Spidstype:
    LTS compatible, extended length
  • Spits farge:
    Clear
  • Filter:
    -