Orders LinkContactUsLinkComponent
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
LTS compatible, 200 µl, +, -, 50,29 mm, Clear, 10 racks, 96 each
Katalog # 613-7416
Leverandør:   Corning
Axygen® LTS Compatible Pipette Tips
Katalog # 613-7416
Leverandør:  Corning
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips
LTS compatible, 200 µl, +, -, 50,29 mm, Clear, 10 racks, 96 each
Price
498,00 kr
Log ind for tilgængelighed
1 * 960 Items
Product Details & Documents
The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.
  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing
Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.
 : ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.
Specifikationer
  • Pakket:
    10 racks, 96 each
  • Steril:
    +
  • Volumen:
    200 µl
  • Længde:
    50,29 mm
  • Seq:
    05
  • Spidstype:
    LTS compatible
  • Spits farge:
    Clear
  • Filter:
    -