Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Axygen® LTS Compatible Pipette Tips

Axygen® LTS Compatible Pipette Tips

LTS compatible, 200 µl, -, -, 50,29 mm, Clear, 10 racks, 96 each
Katalog # 613-7417
Leverandør:   Corning
Axygen® LTS Compatible Pipette Tips
Katalog # 613-7417
Leverandør:  Corning
Axygen® LTS Compatible Pipette Tips
Axygen® LTS Compatible Pipette Tips

LTS compatible, 200 µl, -, -, 50,29 mm, Clear, 10 racks, 96 each

Price
344,00 kr
Log ind for tilgængelighed
1 * 960 Items
Product Details & Documents

The new Axygen® LTS compatible pipette tips are ergonomically designed to reduce discomfort and are optimised to work with Rainin LiteTouch™ pipettors. These tips maintain the quality users have come to expect from corning.

  • Graduation lines for quick accuracy checks
  • Ultra-clear USP class VI resin
  • Fine point delivery system ensures crisp sample dispensing

Flexible sealing area optimised to work with LTS pipettors enables less force to be used during pipetting.

Certifications: ISO 13485, ISO 14001. RNase-free, DNase-free, human DNA-free, PCR inhibitor-free, and non-pyrogenic.

Specifikationer
  • Pakket:
    10 racks, 96 each
  • Steril:
    -
  • Volumen:
    200 µl
  • Længde:
    50,29 mm
  • Seq:
    11
  • Spidstype:
    LTS compatible
  • Spits farge:
    Clear
  • Filter:
    -
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.