Aspartame
Poly pail, 500 g
Katalog # SPCMA1378-500GM
Leverandør: Spectrum Chemical
Gå til:
Product Details & Documents
Aspartame, Powder, NF. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
- Transmittance at 430 nm: minimum 0,95
- Specific rotation (A)D: +14,5 to +16,5
- Loss On drying: Max. 4,5%
- Residue on ignition: Max. 0,2%
Specifikationer
Specifikationer
- Pakning:Poly pail
- Pakn:500 g
- Kvalitet:NF
- Cat. No.:SPCMA1378-500GM
- CAS-nr:22839-47-0
- Molecular formula:C₁₄H₁₈N₂O₅
- Form:Powder
- Protein/peptide name:Aspartame
- Protein synonyms:Methyl aspartylphenylalanate|stereoisomer|L-Phenylalanine, N-L-alpha-aspartyl, 1-methyl ester|N-L-alpha Aspartyl-L-phenlalanine 1-methyl ester|Succinamic acid|Nutrasweet|N-methyl ester|3-amino-N-(alpha-carboxyphenethyl)-|1-Methyl N-L-alpha-aspartyl-L-phenylalanine|Aspartylphenylalanine methyl ester|3-Amino-N-(alpha-carboxyphenethyl) succinamic acid N-methyl ester
- Molekylvægt:294,30 g/mol
- MDL No.:MFCD00002724
- Renhed:98,0% (Dried basis)
- Opbevaring betingelser:Ambient
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



