Membrane Filters, Nylon
Membrane filters, nylon, white, Plain disks, 5,00 µm, 13 mm, +
Katalog # ADVTN500A013A
Leverandør: Advantech
Gå til:
Product Details & Documents
These membrane filters are inherently hydrophilic membranes made by impregnating a polyester web with nylon and are used for HPLC sample preparation, to sterilize biological solutions or buffers, or for vacuum degassing.
- High strength and heat resistance
- Binds proteins, DNA and RNA
- Pure, negligible organic extractables
- Maximum operating temperature: 180 °C
- Compatible with many aqueous and alcoholic solvents and solutions
Suitable for HPLC sample preparation. Filter sterilise and clarify aqueous and organic solvent solutions including buffers, microbiological and tissue culture solutions. Vacuum degassing.
Specifikationer
- Ø:13 mm
- Porestørrelse:5,00 µm
- Steril:+
- Model:Plain disks
- Beskrivelse:Membrane filters, nylon, white
- Seq:01
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



