Orders LinkContactUsLinkComponent
Accessories for Cryo Boxes
Accessories for Cryo Boxes
Divider, PP, natural, 10×10 compartments, 1×121×27 mm, 36 g
Katalog # GLWAZP100
Leverandør:   GLW
Accessories for Cryo Boxes
Katalog # GLWAZP100
Leverandør:  GLW
Accessories for Cryo Boxes
Accessories for Cryo Boxes
Divider, PP, natural, 10×10 compartments, 1×121×27 mm, 36 g
Price
68,50 kr
Log ind for tilgængelighed
1 * 18 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    Divider, PP, natural, 10×10 compartments, 1×121×27 mm, 36 g
  • Seq:
    14